Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003813.3 | NP_003804.2 | 2 | 712 | ||
GenBank mRNA | BC109024.1 | - | 1 | 454 | 40027108 | |
BC109025.1 | - | 1 | 453 |
Gene Symbol: ADAM21P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003951.1 | 1 | 595 | |||
GenBank mRNA | DQ202398.1 | - | 1 | 595 |