Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320336.1 | NP_001307265.1 | 5 | 1757 | ||
NM_005099.5 | NP_005090.3 | 5 | 1757 | |||
GenBank mRNA | AB014588.1 | - | 5 | 1731 | ||
AF148213.1 | - | 5 | 1737 | |||
AK291573.1 | - | 5 | 1756 | |||
AY358886.1 | - | 5 | 1749 | |||
BC063293.1 | - | 5 | 1756 | 5303796 |