Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144822.1 | NP_001138294.1 | 2 | 391 | ||
NM_001779.2 | NP_001770.1 | 2 | 391 | |||
NR_026665.1 | 2 | 391 | ||||
XM_017002869.1 | XP_016858358.1 | 2 | 391 | |||
GenBank mRNA | AK291980.1 | - | 2 | 324 | ||
BC005930.1 | - | 2 | 337 | 4285827 | ||
BT019816.1 | - | 2 | 270 | |||
BT019817.1 | - | 2 | 270 | |||
CR456939.1 | - | 2 | 270 | |||
D28586.1 | - | 1 | 186 | |||
S79616.1 | - | 1 | 162 | |||
X06296.1 | - | 2 | 282 | |||
Y00636.1 | - | 2 | 279 |