Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135703.2 | NP_001129175.1 | 2 | 320 | ||
NM_013437.4 | NP_038465.1 | 3 | 377 | |||
GenBank mRNA | AB208957.1 | - | 1 | 52 | ||
AF064841.1 | - | 2 | 312 | |||
AF166350.1 | - | 3 | 292 | |||
AK289752.1 | - | 3 | 281 | |||
AK299374.1 | - | 2 | 317 | |||
AL698796.1 | - | 3 | 345 | |||
BC032109.2 | - | 3 | 253 | 4842797 | ||
CR749434.1 | - | 2 | 292 | |||
DC372924.1 | - | 2 | 317 |