Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMPCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282944.1 | NP_001269873.1 | 6 | 855 | ||
NM_001282946.1 | NP_001269875.1 | 7 | 948 | |||
NM_015160.2 | NP_055975.1 | 7 | 950 | |||
XM_005266059.4 | XP_005266116.1 | 7 | 865 | |||
XM_011518417.2 | XP_011516719.1 | 3 | 404 | |||
XM_017014543.1 | XP_016870032.1 | 6 | 811 | |||
GenBank mRNA | AK091455.1 | - | 1 | 1105 | ||
AK296617.1 | - | 6 | 767 | |||
AK308104.1 | - | 7 | 863 | |||
BC022949.1 | - | 7 | 842 | |||
BC033103.1 | - | 7 | 859 | |||
BC111399.1 | - | 7 | 865 | 4609491 | ||
BC132724.1 | - | 7 | 889 | 40146746 | ||
BC136599.1 | - | 7 | 889 | 9020589 | ||
D21064.1 | - | 7 | 863 | |||
D50913.2 | - | 7 | 863 |