Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADAM22 SIRNA Assays ›
Gene Symbol:
ADAM22
Gene Name:
ADAM metallopeptidase domain 22
Gene Aliases:
ADAM 22, MDC2
Chromosome Location:
Chr.7: 87934143 - 88202889 on Build GRCh38
UniGene ID:
Hs.256398
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
104159
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADAM22
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001324417.1
NP_001311346.1
3
604
NM_001324418.1
NP_001311347.1
3
607
NM_001324419.1
NP_001311348.1
3
604
NM_001324420.1
NP_001311349.1
3
607
NM_001324421.1
NP_001311350.1
3
607
NM_004194.4
NP_004185.1
3
607
NM_016351.5
NP_057435.2
3
607
NM_021721.4
NP_068367.1
3
607
NM_021722.5
NP_068368.2
3
607
NM_021723.4
NP_068369.1
3
607
XM_005250445.3
XP_005250502.1
2
455
XM_006716028.3
XP_006716091.1
2
455
XM_006716029.3
XP_006716092.1
2
455
XM_011516318.1
XP_011514620.1
3
499
XM_011516319.2
XP_011514621.1
3
583
XM_011516320.2
XP_011514622.1
2
455
XM_011516321.2
XP_011514623.1
2
455
XM_011516322.1
XP_011514624.1
3
499
XM_011516323.1
XP_011514625.1
3
499
XM_011516324.1
XP_011514626.1
3
171
XM_017012329.1
XP_016867818.1
4
550
XM_017012330.1
XP_016867819.1
4
550
XM_017012331.1
XP_016867820.1
3
580
XM_017012332.1
XP_016867821.1
3
499
XM_017012333.1
XP_016867822.1
3
499
XM_017012334.1
XP_016867823.1
4
550
XM_017012335.1
XP_016867824.1
4
550
XM_017012336.1
XP_016867825.1
3
499
XM_017012337.1
XP_016867826.1
4
550
XM_017012338.1
XP_016867827.1
3
578
XM_017012339.1
XP_016867828.1
4
550
XM_017012341.1
XP_016867830.1
2
196
GenBank mRNA
AB009671.1
-
3
405
AF073291.2
-
3
330
AF155381.1
-
3
330
AF155382.1
-
3
330
AF158637.1
-
2
167
AK125432.1
-
2
470
AK291010.1
-
3
454
BC036029.1
-
3
499
BC062433.1
-
3
438
BC092465.1
-
3
499
BC125118.1
-
3
361
40118904
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
cell adhesion
negative regulation of cell adhesion
central nervous system development
adult locomotory behavior
myelination in peripheral nervous system
metalloendopeptidase activity
integrin binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0