Search Thermo Fisher Scientific
Gene Symbol: ADAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033274.4 | NP_150377.1 | 12 | 1330 | ||
XM_005266003.2 | XP_005266060.1 | 12 | 1282 | |||
XM_011534682.2 | XP_011532984.1 | 5 | 2086 | |||
XM_017010009.1 | XP_016865498.1 | 5 | 547 | |||
GenBank mRNA | AF134707.1 | - | 9 | 911 | ||
AF311317.1 | - | 12 | 1239 | |||
AF326918.1 | - | 12 | 1251 | |||
AK091464.1 | - | 12 | 1292 | |||
AK308182.1 | - | 12 | 1282 | |||
Y13786.2 | - | 12 | 1280 |