Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MMEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033467.3 | NP_258428.2 | 21, 22 | 2289 | ||
XM_011542122.2 | XP_011540424.1 | 14, 15 | 1392 | |||
XM_017002310.1 | XP_016857799.1 | 22, 23 | 2555 | |||
XM_017002311.1 | XP_016857800.1 | 21, 22 | 2235 | |||
XM_017002312.1 | XP_016857801.1 | 20, 21 | 2207 | |||
XM_017002314.1 | XP_016857803.1 | 20, 21 | 2160 | |||
XM_017002315.1 | XP_016857804.1 | 21, 22 | 2165 | |||
GenBank mRNA | AF336981.1 | - | 20, 21 | 2112 | ||
AK093058.1 | - | 21, 22 | 2237 | |||
BC101027.2 | - | 20, 21 | 2085 | 40007698 | ||
BC101028.1 | - | 16, 17 | 1614 | 40007699 | ||
BC101029.2 | - | 20, 21 | 2114 | 40007702 | ||
BC101030.1 | - | 20, 21 | 2085 | 40007706 |