Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326358.1 | NP_001313287.1 | 3 | 865 | ||
NM_199355.3 | NP_955387.1 | 3 | 865 | |||
XM_011522923.1 | XP_011521225.1 | 1 | 272 | |||
XM_011522924.1 | XP_011521226.1 | 1 | 268 | |||
XM_017022988.1 | XP_016878477.1 | 1 | 268 | |||
XM_017022989.1 | XP_016878478.1 | 1 | 69 | |||
GenBank mRNA | AJ311903.1 | - | 3 | 528 | ||
AK131403.1 | - | 2 | 372 | |||
AK293831.1 | - | 3 | 785 | |||
BC063283.1 | - | 3 | 865 | 30343625 |