Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139056.3 | NP_620687.2 | 3, 4 | 642 | ||
NR_136935.1 | 3, 4 | 642 | ||||
GenBank mRNA | AJ315734.1 | - | 3, 4 | 504 | ||
AK122980.1 | - | 1, 2 | 267 | |||
AK127746.1 | - | 3, 4 | 642 | |||
DQ266047.1 | - | 3, 4 | 642 |