Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139057.3 | NP_620688.2 | 20 | 2971 | ||
XM_005254872.3 | XP_005254929.1 | 21 | 3968 | |||
XM_011521312.2 | XP_011519614.1 | 22 | 4037 | |||
XM_017021973.1 | XP_016877462.1 | 23 | 4170 | |||
XM_017021974.1 | XP_016877463.1 | 23 | 4172 | |||
XM_017021975.1 | XP_016877464.1 | 22 | 4101 | |||
XM_017021976.1 | XP_016877465.1 | 22 | 2882 | |||
XM_017021978.1 | XP_016877467.1 | 16 | 5428 | |||
XM_017021979.1 | XP_016877468.1 | 15 | 1878 | |||
XM_017021980.1 | XP_016877469.1 | 15 | 2088 | |||
XM_017021982.1 | XP_016877471.1 | 13 | 1698 | |||
XM_017021983.1 | XP_016877472.1 | 11 | 1427 | |||
GenBank mRNA | AJ315735.1 | - | 20 | 2888 |