Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006690.3 | NP_006681.1 | 9 | 2553 | ||
XM_011528500.2 | XP_011526802.1 | 8 | 2283 | |||
XM_017027597.1 | XP_016883086.1 | 8 | 2388 | |||
XM_017027598.1 | XP_016883087.1 | 9 | 2524 | |||
GenBank mRNA | AB021227.1 | - | 9 | 2552 | ||
BC047614.1 | - | 9 | 2493 |