Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: METAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015143.2 | NP_055958.2 | 11 | 1407 | ||
XM_011531777.2 | XP_011530079.1 | 11 | 1404 | |||
XM_011531778.1 | XP_011530080.1 | 11 | 1200 | |||
XM_011531779.1 | XP_011530081.1 | 10 | 1275 | |||
XM_017007914.1 | XP_016863403.1 | 10 | 1263 | |||
XM_017007915.1 | XP_016863404.1 | 12 | 1421 | |||
XM_017007916.1 | XP_016863405.1 | 11 | 1420 | |||
XM_017007917.1 | XP_016863406.1 | 11 | 1196 | |||
GenBank mRNA | AK304239.1 | - | 11 | 1323 | ||
AK309465.1 | - | 3 | 480 | |||
BC030054.1 | - | 7 | 965 | 3958447 | ||
D42084.1 | - | 11 | 1315 |