Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: METAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015143.2 | NP_055958.2 | 11 | 2234 | ||
XM_011531777.2 | XP_011530079.1 | 11 | 2231 | |||
XM_011531778.1 | XP_011530080.1 | 11 | 2027 | |||
XM_011531779.1 | XP_011530081.1 | 10 | 2102 | |||
XM_017007914.1 | XP_016863403.1 | 10 | 2090 | |||
XM_017007915.1 | XP_016863404.1 | 12 | 2248 | |||
XM_017007916.1 | XP_016863405.1 | 11 | 2247 | |||
XM_017007917.1 | XP_016863406.1 | 11 | 2023 | |||
GenBank mRNA | BC030054.1 | - | 7 | 1792 | 3958447 | |
D42084.1 | - | 11 | 2142 |