Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256959.1 | NP_001243888.1 | 22 | 4562 | ||
NM_001286.3 | NP_001277.1 | 23 | 4628 | |||
NR_046428.1 | 23 | 4612 | ||||
GenBank mRNA | D28475.2 | - | 23 | 4513 | ||
X83378.1 | - | 23 | 4487 |