Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001567.3 | NP_001558.3 | 8, 9 | 1094 | ||
XM_005273978.3 | XP_005274035.1 | 9, 10 | 1471 | |||
XM_005273979.4 | XP_005274036.1 | 9, 10 | 1318 | |||
XM_011544999.2 | XP_011543301.1 | 9, 10 | 1252 | |||
XM_011545000.2 | XP_011543302.1 | 9, 10 | 1318 | |||
XM_017017673.1 | XP_016873162.1 | 10, 11 | 1409 | |||
GenBank mRNA | BC140853.1 | - | 8, 9 | 1094 | 9021721 | |
Y14385.1 | - | 8, 9 | 1158 |