Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002709.2 | NP_002700.1 | 8 | 2505 | ||
NM_206876.1 | NP_996759.1 | 9 | 2300 | |||
GenBank mRNA | BC002697.2 | - | 8 | 2220 | 3609386 | |
BC012045.1 | - | 8 | 2220 | 3631338 | ||
BX647295.1 | - | 4 | 1696 | |||
BX647970.1 | - | 9 | 2338 | |||
EU668339.1 | - | 8 | 2220 | |||
X80910.1 | - | 8 | 2398 |