Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPP2CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002715.2 | NP_002706.1 | 2, 3 | 718 | ||
GenBank mRNA | AB451240.1 | - | 2, 3 | 320 | ||
AB451365.1 | - | 2, 3 | 320 | |||
AK057424.1 | - | 2, 3 | 450 | |||
AK097599.1 | - | 2, 3 | 530 | |||
BC000400.2 | - | 2, 3 | 519 | 2822635 | ||
BC002657.2 | - | 2, 3 | 519 | 3607862 | ||
BC019275.1 | - | 2, 3 | 519 | 2822635 | ||
BC031696.1 | - | 2, 3 | 519 | 5001649 | ||
CR457417.1 | - | 2, 3 | 320 | |||
J03804.1 | - | 2, 3 | 502 | |||
X12646.1 | - | 2, 3 | 403 |