Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPP2CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002715.2 | NP_002706.1 | 7 | 1768 | ||
GenBank mRNA | AK057424.1 | - | 7 | 1500 | ||
AK097599.1 | - | 7 | 1580 | |||
BC000400.2 | - | 7 | 1569 | 2822635 | ||
BC002657.2 | - | 7 | 1569 | 3607862 | ||
BC019275.1 | - | 7 | 1569 | 2822635 | ||
BC031696.1 | - | 7 | 1569 | 5001649 | ||
J03804.1 | - | 7 | 1551 |