Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195100.1 | NP_001182029.1 | 4 | 1048 | ||
NM_001195101.1 | NP_001182030.1 | 4 | 1030 | |||
NM_080391.3 | NP_536316.1 | 6 | 1524 | |||
XM_005271231.3 | XP_005271288.1 | 5 | 1151 | |||
XM_005271232.3 | XP_005271289.1 | 5 | 1133 | |||
XM_017002401.1 | XP_016857890.1 | 5 | 1066 | |||
GenBank mRNA | AF208850.1 | - | 5 | 871 | ||
AK129578.1 | - | 2 | 1380 | |||
AK292703.1 | - | 5 | 632 | |||
AK297280.1 | - | 4 | 740 | |||
BC018662.1 | - | 6 | 1495 | |||
BC070182.1 | - | 5 | 858 | 4686913 | ||
L39000.1 | - | 5 | 493 | |||
U14603.1 | - | 5 | 953 | |||
U48297.1 | - | 5 | 595 |