Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040151.1 | NP_001035241.1 | 7 | 1497 | ||
NM_018400.3 | NP_060870.1 | 6 | 1896 | |||
GenBank mRNA | AB032984.1 | - | 7 | 1137 | ||
AL136589.1 | - | 6 | 1893 | |||
BC117282.1 | - | 6 | 1183 | 40125833 | ||
BC126265.1 | - | 6 | 1183 | 8991981 |