Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014437.2 | NP_001014437.1 | 23 | 2598 | ||
NM_001194997.1 | NP_001181926.1 | 23 | 2606 | |||
NM_001751.5 | NP_001742.1 | 22 | 2349 | |||
NM_139273.3 | NP_644802.1 | 22 | 2357 | |||
NR_036542.1 | 23 | 2645 | ||||
XM_006718341.3 | XP_006718404.1 | 22 | 2372 | |||
XM_017018389.1 | XP_016873878.1 | 22 | 2380 | |||
XM_017018390.1 | XP_016873879.1 | 23 | 2620 | |||
XM_017018391.1 | XP_016873880.1 | 20 | 2596 | |||
XM_017018392.1 | XP_016873881.1 | 22 | 2542 | |||
GenBank mRNA | AB208814.1 | - | 19 | 5566 | ||
AF288206.1 | - | 22 | 2282 | |||
AF288207.1 | - | 22 | 2290 | |||
AK096313.1 | - | 23 | 2612 | |||
AK125503.1 | - | 5 | 2505 | |||
AK126135.1 | - | 5 | 2288 | |||
AK222614.1 | - | 22 | 2298 | |||
AK295371.1 | - | 22 | 2248 | |||
AK296763.1 | - | 23 | 2471 | |||
AK298514.1 | - | 23 | 2573 | |||
AK302644.1 | - | 23 | 2565 | |||
BC002880.2 | - | 22 | 2335 | 3940167 | ||
BX647906.1 | - | 23 | 2572 | |||
L06845.1 | - | 20 | 2142 |