Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297741.1 | NP_001284670.1 | 2 | 310 | ||
NM_001297742.1 | NP_001284671.1 | 2 | 500 | |||
NM_032320.6 | NP_115696.2 | 3 | 468 | |||
XM_017018405.1 | XP_016873894.1 | 2 | 1479 | |||
XM_017018406.1 | XP_016873895.1 | 3 | 458 | |||
XM_017018407.1 | XP_016873896.1 | 3 | 438 | |||
XM_017018408.1 | XP_016873897.1 | 3 | 438 | |||
XM_017018409.1 | XP_016873898.1 | 2 | 305 | |||
XM_017018410.1 | XP_016873899.1 | 2 | 297 | |||
XM_017018411.1 | XP_016873900.1 | 2 | 297 | |||
GenBank mRNA | AF329102.2 | - | 3 | 468 | ||
AK095946.1 | - | 3 | 397 | |||
AK292392.1 | - | 3 | 393 | |||
AK294259.1 | - | 2 | 289 | |||
AK294768.1 | - | 2 | 215 | |||
AK298226.1 | - | 2 | 185 | |||
AK310935.1 | - | 2 | 193 | |||
AK316163.1 | - | 2 | 500 | |||
AL832981.1 | - | 3 | 449 | |||
AY221959.2 | - | 3 | 511 | |||
BC005071.1 | - | 3 | 430 | 3531347 |