Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142370.1 | NP_001135842.1 | 11 | 1353 | ||
NM_014369.3 | NP_055184.2 | 15 | 1674 | |||
GenBank mRNA | AK297884.1 | - | 3 | 748 | ||
AK303804.1 | - | 11 | 1276 | |||
AK311140.1 | - | 2 | 490 | |||
BC024280.1 | - | 11 | 1309 | |||
BC041562.1 | - | 11 | 1309 | |||
BC052800.1 | - | 11 | 1309 | 5809095 | ||
X79568.1 | - | 15 | 1609 |