Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPM1A SIRNA Assays ›
Gene Symbol:
PPM1A
Gene Name:
protein phosphatase, Mg2+/Mn2+ dependent 1A
Gene Aliases:
PP2C-ALPHA, PP2CA, PP2Calpha
Chromosome Location:
Chr.14: 60245752 - 60299087 on Build GRCh38
UniGene ID:
Hs.130036
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
104943
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPM1A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_021003.4
NP_066283.1
2
993
NM_177951.2
NP_808820.1
3
1095
NM_177952.2
NP_808821.2
2
858
XM_005267779.1
XP_005267836.1
3
940
XM_005267781.1
XP_005267838.1
3
1098
XM_011536872.1
XP_011535174.1
5
1360
XM_011536876.1
XP_011535178.1
4
1217
XM_011536877.1
XP_011535179.1
5
1260
XM_011536878.1
XP_011535180.1
4
1183
XM_011536879.2
XP_011535181.1
4
1052
XM_011536880.2
XP_011535182.1
5
1106
XM_011536881.2
XP_011535183.1
3
966
XM_011536882.2
XP_011535184.1
2
635
XM_011536883.1
XP_011535185.1
3
1049
XM_011536884.1
XP_011535186.1
2
606
XM_017021381.1
XP_016876870.1
5
2230
XM_017021382.1
XP_016876871.1
7
1480
XM_017021383.1
XP_016876872.1
4
1116
XM_017021384.1
XP_016876873.1
4
808
XM_017021385.1
XP_016876874.1
3
1083
XM_017021386.1
XP_016876875.1
3
846
XM_017021387.1
XP_016876876.1
4
1092
GenBank mRNA
AB451247.1
-
1
543
AB451371.1
-
1
543
AF070670.1
-
3
902
AK097843.1
-
2
858
AK308225.1
-
2
720
AK313337.1
-
3
940
BC026691.1
-
2
846
3847638
BC063243.1
-
2
776
6011486
S87759.1
-
2
900
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
protein dephosphorylation
N-terminal protein myristoylation
cell cycle arrest
negative regulation of SMAD protein complex assembly
Wnt signaling pathway
dephosphorylation
positive regulation of Wnt signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of BMP signaling pathway
peptidyl-threonine dephosphorylation
negative regulation of NF-kappaB import into nucleus
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of transcription, DNA-templated
positive regulation of protein export from nucleus
cellular response to transforming growth factor beta stimulus
magnesium ion binding
protein serine/threonine phosphatase activity
signal transducer activity
protein binding
manganese ion binding
calmodulin-dependent protein phosphatase activity
R-SMAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline