Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP3CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142353.2 | NP_001135825.1 | 14 | 2171 | ||
NM_001142354.2 | NP_001135826.1 | 13 | 2141 | |||
NM_021132.3 | NP_066955.1 | 14 | 2168 | |||
XM_005269944.1 | XP_005270001.1 | 13 | 2138 | |||
XM_017016379.1 | XP_016871868.1 | 13 | 2073 | |||
GenBank mRNA | AJ488506.1 | - | 13 | 2078 | ||
AK300701.1 | - | 6 | 964 | |||
BC028049.1 | - | 14 | 2159 | 4538822 | ||
BC035464.1 | - | 14 | 2126 | |||
M29551.1 | - | 14 | 2149 |