Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PTPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199797.1 | NP_001186726.1 | 6 | 816 | ||
NM_002832.3 | NP_002823.3 | 6 | 1662 | |||
NM_080588.2 | NP_542155.1 | 5 | 1155 | |||
NR_037663.1 | 6 | 1609 | ||||
NR_037664.1 | 5 | 1478 | ||||
XM_011509819.1 | XP_011508121.1 | 6 | 997 | |||
XM_011509820.2 | XP_011508122.1 | 6 | 701 | |||
XM_011509821.1 | XP_011508123.1 | 6 | 629 | |||
XM_011509822.2 | XP_011508124.1 | 6 | 500 | |||
XM_011509823.1 | XP_011508125.1 | 6 | 997 | |||
XM_011509824.1 | XP_011508126.1 | 5 | 505 | |||
XM_017001935.1 | XP_016857424.1 | 6 | 994 | |||
XM_017001936.1 | XP_016857425.1 | 6 | 994 | |||
XM_017001937.1 | XP_016857426.1 | 6 | 812 | |||
GenBank mRNA | AF394064.1 | - | 4 | 599 | ||
AK127214.1 | - | 6 | 1655 | |||
AK225920.1 | - | 6 | 660 | |||
AK293186.1 | - | 5 | 505 | |||
AK301051.1 | - | 6 | 630 | |||
AK302869.1 | - | 6 | 810 | |||
BC001746.1 | - | 5 | 1155 | 3543111 | ||
BT009848.1 | - | 5 | 566 | |||
D11327.1 | - | 6 | 670 | |||
M64322.1 | - | 6 | 658 |