- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Gene Symbol: PPP2R2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271899.1 | NP_001258828.1 | 8 | 1161 | ||
NM_001271900.1 | NP_001258829.1 | 9 | 1355 | |||
NM_001271948.1 | NP_001258877.1 | 8 | 1688 | |||
NM_181674.2 | NP_858060.2 | 8 | 1486 | |||
NM_181675.3 | NP_858061.2 | 8 | 1208 | |||
NM_181676.2 | NP_858062.1 | 7 | 1292 | |||
NM_181677.2 | NP_858063.1 | 9 | 1118 | |||
NM_181678.2 | NP_858064.1 | 9 | 1131 | |||
NR_073526.1 | 7 | 1603 | ||||
NR_073527.1 | 8 | 1240 | ||||
GenBank mRNA | AK056192.1 | - | 8 | 1168 | ||
AK289717.1 | - | 8 | 1137 | |||
AK294659.1 | - | 9 | 1264 | |||
AK295347.1 | - | 8 | 1132 | |||
AK314810.1 | - | 8 | 1485 | |||
BC031790.1 | - | 8 | 1376 | 4939981 | ||
BX647887.1 | - | 8 | 1222 | |||
M64930.1 | - | 7 | 1414 |