Gene Symbol: PPP2R2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271899.1 | NP_001258828.1 | 8 | 1176 | ||
NM_001271900.1 | NP_001258829.1 | 9 | 1370 | |||
NM_001271948.1 | NP_001258877.1 | 8 | 1703 | |||
NM_181674.2 | NP_858060.2 | 8 | 1501 | |||
NM_181675.3 | NP_858061.2 | 8 | 1223 | |||
NM_181676.2 | NP_858062.1 | 7 | 1307 | |||
NM_181677.2 | NP_858063.1 | 9 | 1133 | |||
NM_181678.2 | NP_858064.1 | 9 | 1146 | |||
NR_073526.1 | 7 | 1618 | ||||
NR_073527.1 | 8 | 1255 | ||||
GenBank mRNA | AK056192.1 | - | 8 | 1183 | ||
AK289717.1 | - | 8 | 1152 | |||
AK294659.1 | - | 9 | 1279 | |||
AK295347.1 | - | 8 | 1147 | |||
AK314810.1 | - | 8 | 1500 | |||
BC031790.1 | - | 8 | 1391 | 4939981 | ||
BX647887.1 | - | 8 | 1237 | |||
M64930.1 | - | 7 | 1429 |