Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256702.1 | NP_001243631.1 | 14 | 2195 | ||
NM_006537.3 | NP_006528.2 | 15 | 2327 | |||
NR_046341.1 | 16 | 2480 | ||||
NR_046342.1 | 16 | 2443 | ||||
XM_017022763.1 | XP_016878252.1 | 17 | 3326 | |||
XM_017022764.1 | XP_016878253.1 | 16 | 3262 | |||
XM_017022765.1 | XP_016878254.1 | 16 | 2468 | |||
XM_017022766.1 | XP_016878255.1 | 17 | 2511 | |||
XM_017022767.1 | XP_016878256.1 | 18 | 3442 | |||
XM_017022768.1 | XP_016878257.1 | 18 | 3516 | |||
GenBank mRNA | AF073344.1 | - | 15 | 2195 | ||
AF077040.1 | - | 14 | 1973 | |||
AK022931.1 | - | 13 | 1913 | |||
AK307810.1 | - | 15 | 2086 | |||
AY461579.1 | - | 16 | 2356 | |||
BC018113.2 | - | 15 | 2192 | 3916811 | ||
BC065300.1 | - | 15 | 2227 | 6146202 | ||
BX647760.1 | - | 16 | 2221 |