Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014296.2 | NP_055111.1 | 20 | 2554 | ||
XM_011533555.2 | XP_011531857.1 | 21 | 2453 | |||
XM_011533556.2 | XP_011531858.1 | 22 | 2685 | |||
XM_011533557.2 | XP_011531859.1 | 19 | 2293 | |||
XM_017006043.1 | XP_016861532.1 | 21 | 2637 | |||
XM_017006044.1 | XP_016861533.1 | 18 | 2245 | |||
XM_017006045.1 | XP_016861534.1 | 19 | 2311 | |||
GenBank mRNA | AB028639.1 | - | 20 | 2535 | ||
AK093742.1 | - | 3 | 1119 | |||
AK314253.1 | - | 20 | 2516 | |||
BC056202.1 | - | 20 | 2532 | 5274889 |