Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021928.3 | NP_068747.1 | 5 | 630 | ||
GenBank mRNA | AK026302.1 | - | 5 | 605 | ||
AK092634.1 | - | 4 | 1613 | |||
AL136660.1 | - | 5 | 600 | |||
AY359044.1 | - | 5 | 588 | |||
BC047290.1 | - | 5 | 573 | 5224927 | ||
BC053883.1 | - | 4 | 2524 |