Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293557.1 | NP_001280486.1 | 11 | 3131 | ||
NM_022162.2 | NP_071445.1 | 12 | 3171 | |||
XM_005256084.3 | XP_005256141.1 | 12 | 3075 | |||
XM_006721242.3 | XP_006721305.1 | 11 | 2991 | |||
XM_011523259.2 | XP_011521561.1 | 10 | 2887 | |||
XM_017023535.1 | XP_016879024.1 | 10 | 4445 | |||
XM_017023536.1 | XP_016879025.1 | 11 | 2602 | |||
XM_017023537.1 | XP_016879026.1 | 10 | 2496 | |||
XM_017023538.1 | XP_016879027.1 | 11 | 2597 | |||
GenBank mRNA | AF178930.1 | - | 12 | 3171 | ||
AK298099.1 | - | 7 | 746 |