Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UQCRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003365.2 | NP_003356.2 | 4 | 441 | ||
GenBank mRNA | AK129903.1 | - | 4 | 421 | ||
AK300694.1 | - | 4 | 376 | |||
AK302840.1 | - | 4 | 466 | |||
AK313090.1 | - | 4 | 411 | |||
BC009586.1 | - | 4 | 412 | 4096628 | ||
D26485.1 | - | 4 | 403 | |||
L16842.1 | - | 4 | 813 |