Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018646.5 | NP_061116.5 | 3, 4 | 576 | ||
GenBank mRNA | AF304463.1 | - | 3, 4 | 347 | ||
AF365927.1 | - | 3, 4 | 563 | |||
AJ243500.2 | - | 3, 4 | 547 | |||
AJ243501.2 | - | 3, 4 | 547 | |||
AJ487964.2 | - | 4, 5 | 516 | |||
AK291707.1 | - | 3, 4 | 577 | |||
KF534785.1 | - | 3, 4 | 576 |