Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167576.1 | NP_001161048.1 | 2 | 967 | ||
NM_001167577.1 | NP_001161049.1 | 2 | 967 | |||
NM_020389.2 | NP_065122.1 | 2 | 967 | |||
GenBank mRNA | AJ272034.1 | - | 1 | 745 | ||
AJ421783.1 | - | 1 | 745 | |||
AJ549088.1 | - | 1 | 745 | |||
AJ549089.1 | - | 1 | 745 | |||
AJ549090.1 | - | 1 | 745 | |||
AY167926.1 | - | 1 | 745 | |||
AY167927.1 | - | 1 | 745 | |||
BC128185.1 | - | 1 | 745 | 40113370 | ||
BC128186.1 | - | 1 | 745 |