Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001226.3 | NP_001217.2 | 5 | 527 | ||
NM_032992.2 | NP_116787.1 | 2 | 260 | |||
NR_133012.1 | 4 | 450 | ||||
GenBank mRNA | BC000305.2 | - | 5 | 475 | 2820468 | |
BC004460.1 | - | 5 | 475 | 2820468 | ||
BM837197.1 | - | 2 | 206 | |||
BX380795.2 | - | 4 | 391 | |||
U20536.1 | - | 5 | 527 | |||
U20537.1 | - | 2 | 182 |