Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002775.4 | NP_002766.1 | 2 | 617 | ||
GenBank mRNA | AF097709.1 | - | 2 | 60 | ||
AK091944.1 | - | 4 | 392 | |||
AK092476.1 | - | 3 | 398 | |||
BC011352.1 | - | 3 | 384 | |||
BC031082.1 | - | 2 | 525 | |||
D87258.1 | - | 2 | 537 | |||
DA633502.1 | - | 4 | 392 | |||
Y07921.1 | - | 2 | 527 |