Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282669.1 | NP_001269598.1 | 4 | 855 | ||
NM_001320132.1 | NP_001307061.1 | 4 | 952 | |||
NM_001320136.1 | NP_001307065.1 | 4 | 833 | |||
NM_004402.3 | NP_004393.1 | 4 | 783 | |||
NR_104222.1 | 5 | 920 | ||||
NR_135150.1 | 4 | 783 | ||||
NR_135151.1 | 5 | 870 | ||||
NR_135152.1 | 4 | 805 | ||||
XM_017000498.1 | XP_016855987.1 | 3 | 718 | |||
XM_017000499.1 | XP_016855988.1 | 4 | 835 | |||
XM_017000500.1 | XP_016855989.1 | 4 | 590 | |||
GenBank mRNA | AB013918.1 | - | 4 | 446 | ||
AB028911.1 | - | 4 | 577 | |||
AB028912.1 | - | 4 | 746 | |||
AB028913.1 | - | 5 | 664 | |||
AF039210.1 | - | 4 | 446 | |||
AF064019.1 | - | 4 | 577 | |||
AF409060.1 | - | 6 | 457 | |||
AF409061.1 | - | 4 | 472 | |||
AF409062.1 | - | 4 | 500 | |||
AK290877.1 | - | 4 | 574 | |||
AK303065.1 | - | 4 | 662 | |||
AY438267.1 | - | 4 | 375 | |||
BC032827.1 | - | 5 | 732 | |||
BC048797.1 | - | 4 | 769 | 5204996 |