Gene Symbol: KREMEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253725.1 | NP_001240654.1 | 8 | 1546 | ||
NM_001253726.1 | NP_001240655.1 | 9 | 1625 | |||
NM_024507.3 | NP_078783.1 | 8 | 1663 | |||
NM_172229.2 | NP_757384.1 | 9 | 1742 | |||
GenBank mRNA | AK027669.1 | - | 8 | 1567 | ||
AK075033.1 | - | 8 | 1487 | |||
AK223422.1 | - | 8 | 1435 | |||
AK301953.1 | - | 9 | 1566 | |||
BC003533.1 | - | 8 | 1470 | 3609280 | ||
BC009383.2 | - | 7 | 2027 | 4127918 |