Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182973.1 | NP_892018.1 | 3, 4 | 413 | ||
XM_011527978.2 | XP_011526280.1 | 5, 6 | 776 | |||
GenBank mRNA | AB109390.1 | - | 4, 5 | 530 | ||
AJ488946.1 | - | 3, 4 | 413 | |||
AJ488947.1 | - | 3, 4 | 413 |