Search Thermo Fisher Scientific
Gene Symbol: DUSP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001946.3 | NP_001937.2 | 3 | 2402 | ||
NM_022652.3 | NP_073143.2 | 2 | 1964 | |||
GenBank mRNA | AB013382.1 | - | 3 | 1934 | ||
AB013602.1 | - | 2 | 1496 | |||
AK091753.1 | - | 4 | 2019 | |||
AK222882.1 | - | 2 | 1510 | |||
BC003143.1 | - | 3 | 1930 | 2906126 | ||
BC003562.1 | - | 3 | 1930 | 3547162 | ||
BC005047.1 | - | 3 | 2006 | 3954486 | ||
BC037236.1 | - | 3 | 2029 | 5183133 | ||
X93920.1 | - | 3 | 1933 |