Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRS SIRNA Assays ›
Gene Symbol:
PTPRS
Gene Name:
protein tyrosine phosphatase, receptor type S
Gene Aliases:
PTPSIGMA, R-PTP-S, R-PTP-sigma
Chromosome Location:
Chr.19: 5205503 - 5340803 on Build GRCh38
UniGene ID:
Hs.744928
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
105840
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002850.3
NP_002841.3
4, 5
605
NM_130853.2
NP_570923.2
4, 5
605
NM_130854.2
NP_570924.2
4, 5
605
NM_130855.2
NP_570925.2
4, 5
605
XM_005259600.2
XP_005259657.1
4, 5
573
XM_005259606.2
XP_005259663.1
4, 5
573
XM_005259607.2
XP_005259664.1
4, 5
573
XM_005259609.1
XP_005259666.1
4, 5
572
XM_005259610.1
XP_005259667.1
4, 5
572
XM_011528157.2
XP_011526459.1
3, 4
443
XM_017027065.1
XP_016882554.1
4, 5
573
XM_017027066.1
XP_016882555.1
4, 5
591
XM_017027067.1
XP_016882556.1
4, 5
489
XM_017027068.1
XP_016882557.1
4, 5
573
XM_017027069.1
XP_016882558.1
4, 5
573
XM_017027070.1
XP_016882559.1
4, 5
573
XM_017027071.1
XP_016882560.1
4, 5
573
XM_017027072.1
XP_016882561.1
4, 5
573
XM_017027074.1
XP_016882563.1
4, 5
572
XM_017027075.1
XP_016882564.1
4, 5
572
XM_017027076.1
XP_016882565.1
4, 5
572
GenBank mRNA
AB209333.1
-
4, 5
605
BC104812.1
-
3, 4
445
8143815
BC143286.1
-
3, 4
445
BC143287.1
-
3, 4
445
9051790
U35234.1
-
3, 4
479
U40317.1
-
3, 4
393
U41725.1
-
3, 4
393
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
chemical synaptic transmission
spinal cord development
cerebellum development
hippocampus development
cerebral cortex development
corpus callosum development
peptidyl-tyrosine dephosphorylation
establishment of endothelial intestinal barrier
transmembrane receptor protein tyrosine phosphatase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0