Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204284.1 | NP_001191213.1 | 2 | 290 | ||
NM_006247.3 | NP_006238.1 | 2 | 290 | |||
XM_017026935.1 | XP_016882424.1 | 2 | 285 | |||
GenBank mRNA | AK223222.1 | - | 2 | 197 | ||
AK293402.1 | - | 2, 3 | 187 | |||
AK297095.1 | - | 2 | 187 | |||
AK312683.1 | - | 2 | 197 | |||
BC000750.2 | - | 2 | 183 | |||
BC001831.2 | - | 2 | 183 | |||
BC001970.1 | - | 2 | 247 | 3459309 | ||
BT007275.1 | - | 2 | 187 | |||
DA545760.1 | - | 2 | 290 | |||
U25174.1 | - | 2 | 165 | |||
X89416.1 | - | 2 | 170 |