Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007240.2 | NP_009171.1 | 2 | 401 | ||
XM_005244862.2 | XP_005244919.1 | 2 | 490 | |||
GenBank mRNA | AF119226.1 | - | 2 | 378 | ||
BC006286.1 | - | 2 | 399 | 3958403 | ||
BT006633.1 | - | 2 | 367 | |||
ES309317.1 | - | 2 | 417 | |||
HY128542.1 | - | 2 | 415 |