Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IMPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014214.2 | NP_055029.1 | 2 | 461 | ||
XM_011525661.2 | XP_011523963.1 | 2 | 424 | |||
GenBank mRNA | AF014398.2 | - | 2 | 461 | ||
AF200432.1 | - | 1 | 138 | |||
BC017176.1 | - | 2 | 301 | 3345789 | ||
BC031560.1 | - | 2 | 373 | |||
BT007061.1 | - | 2 | 219 | |||
CF134610.1 | - | 2 | 418 |