Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323625.1 | NP_001310554.1 | 2 | 780 | ||
NM_016361.4 | NP_057445.4 | 2 | 749 | |||
NR_136633.1 | 2 | 780 | ||||
NR_136634.1 | 2 | 1100 | ||||
NR_136635.1 | 2 | 780 | ||||
NR_136636.1 | 2 | 1100 | ||||
XM_011509601.2 | XP_011507903.1 | 2 | 705 | |||
XM_017001422.1 | XP_016856911.1 | 2 | 705 | |||
GenBank mRNA | AB030039.2 | - | 2 | 553 | ||
AB031478.1 | - | 2 | 660 | |||
AB209248.1 | - | 2 | 522 | |||
AK000657.1 | - | 2 | 486 | |||
AL520992.3 | - | 2 | 748 | |||
AY358552.1 | - | 2 | 457 | |||
BC009965.1 | - | 2 | 729 | 4299943 | ||
BC034686.1 | - | 2 | 733 | 3463770 | ||
KJ534759.1 | - | 2 | 285 | |||
KJ535044.1 | - | 2 | 285 | |||
KJ535055.1 | - | 2 | 285 |