Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001917.4 | NP_001908.3 | 4 | 485 | ||
XM_005268692.3 | XP_005268749.1 | 4 | 485 | |||
XM_011538004.2 | XP_011536306.1 | 5 | 571 | |||
XM_011538005.2 | XP_011536307.1 | 5 | 575 | |||
GenBank mRNA | AK312995.1 | - | 4 | 472 | ||
AX741655.1 | - | 4 | 475 | |||
AX741656.1 | - | 4 | 475 | |||
AX741658.1 | - | 3 | 360 | |||
AX776478.1 | - | 4 | 475 | |||
AX776479.1 | - | 4 | 475 | |||
AX776480.1 | - | 7 | 1487 | |||
AX786386.1 | - | 4 | 475 | |||
BC029057.1 | - | 4 | 455 | 5184628 | ||
BC074770.2 | - | 4 | 357 | 30915193 | ||
BX538210.1 | - | 4 | 417 | |||
DA085245.1 | - | 4 | 486 | |||
KU178015.1 | - | 3 | 332 | |||
KU178016.1 | - | 3 | 332 |