Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000018.3 | NP_000009.1 | 10 | 1210 | ||
NM_001033859.2 | NP_001029031.1 | 9 | 1144 | |||
NM_001270447.1 | NP_001257376.1 | 11 | 1146 | |||
NM_001270448.1 | NP_001257377.1 | 9 | 1285 | |||
XM_006721516.2 | XP_006721579.2 | 10 | 1163 | |||
XM_011523829.1 | XP_011522131.1 | 10 | 1163 | |||
XM_011523830.1 | XP_011522132.1 | 10 | 1163 | |||
GenBank mRNA | AK056083.1 | - | 9 | 1178 | ||
AK056956.1 | - | 5 | 556 | |||
AK058109.1 | - | 10 | 1092 | |||
AK097243.1 | - | 8 | 1625 | |||
AK222518.1 | - | 10 | 1092 | |||
AK293549.1 | - | 11 | 1146 | |||
BC000399.2 | - | 10 | 1148 | 2822593 | ||
BC012912.1 | - | 10 | 1132 | 4471040 | ||
BC020218.1 | - | 9 | 1052 | 5090104 | ||
D43682.1 | - | 10 | 1089 | |||
X86556.1 | - | 10 | 1141 |