Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ITGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000212.2 | NP_000203.2 | 15 | 2967 | ||
GenBank mRNA | J02703.1 | - | 15 | 2967 | ||
M20311.1 | - | 15 | 2963 | |||
M35999.1 | - | 15 | 2965 |